1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidin-4-one
047776-500MG
047776-1G
047776-5G
101881-380EA
375.38
USD
101881-380
101881-382
101881-384
1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone ≥95%
1-[4-(Trifluoromethyl)-2-pyrimidinyl]tetrahydro-4(1H)-pyridinone
Hazard: Irritant
Formula:
C₁₀H₁₀F₃N₃O MW: 245.21 g/mol Melting Pt: 110…112 °C |
MDL Number:
MFCD09027162 CAS Number: 937604-44-9 |
Specification Test Results
Melting point | 110-112°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...