tert-Butyl-4-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(tert-Butoxycarbonyl)piperidine-4-carbohydrazide, 1-Boc-isonipecoticacidhydrazide
047774-500MG
047774-1G
047774-5G
101881-368EA
181.54
USD
101881-368
101881-370
101881-372
tert-Butyl-4-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-(hydrazinocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₁H₂₁N₃O₃ MW: 243.31 g/mol Melting Pt: 104…106 °C |
MDL Number:
MFCD06795927 CAS Number: 187834-88-4 |
Specification Test Results
Melting point | 104-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...