N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride ≥95%
Supplier: Matrix Scientific
047768-500MG
047768-1G
101881-334EA
278.46
USD
101881-334
101881-336
N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride ≥95%
N-Hydroxy-2-(trifluoromethyl)benzenecarboximidoylchloride
Hazard: Irritant
Formula:
C₈H₅ClF₃NO Melting Pt: 80…82 °C |
MDL Number:
MFCD09027157 CAS Number: 74467-04-2 |
Specification Test Results
Melting point | 80-82°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...