2-(Trifluoromethyl)benzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

047767-500MG 047767-1G 047767-5G
101881-328EA 181.54 USD
101881-328 101881-330 101881-332
2-(Trifluoromethyl)benzenecarbaldehyde oxime ≥95%
2-(Trifluoromethyl)benzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD03015196
CAS Number: 74467-00-8

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR