2-[4-(Trifluoromethyl)phenyl]-1,3-thiazolane ≥95%

Supplier: Matrix Scientific

047759-500MG 047759-1G 047759-5G
101881-284EA 181.54 USD
101881-284 101881-286 101881-288
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazolane ≥95%
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazolane

Hazard: Irritant

Formula: C₁₀H₁₀F₃NS
Melting Pt: 109…111 °C
MDL Number: MFCD09027152
CAS Number: 937604-42-7

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR