Ethyl-2-(hydroxyimino)-2-(4-phenylpiperazino)acetate ≥95%

Supplier: Matrix Scientific

047756-500MG 047756-1G 047756-5G
101881-272EA 278.46 USD
101881-272 101881-274 101881-276
Ethyl-2-(hydroxyimino)-2-(4-phenylpiperazino)acetate ≥95%
Ethyl-2-(hydroxyimino)-2-(4-phenylpiperazino)acetate

Hazard: Irritant

Formula: C₁₄H₁₉N₃O₃
Melting Pt: 115…117 °C
MDL Number: MFCD09027150
CAS Number: 937604-30-3

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR