Methyl-5-iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridinecarboxylate ≥95%
Supplier: Matrix Scientific
047755-500MG
047755-1G
047755-5G
101881-266EA
556.92
USD
101881-266
101881-268
101881-270
Methyl-5-iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridinecarboxylate ≥95%
Methyl-5-iodo-2-oxo-1-[4-(trifluoromethyl)benzyl]-1,2-dihydro-3-pyridinecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₁F₃INO₃ Melting Pt: 134…136 °C |
MDL Number:
MFCD09027149 CAS Number: 937602-50-1 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...