2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
047754-500MG
047754-1G
047754-5G
101881-260EA
278.46
USD
101881-260
101881-262
101881-264
2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane ≥95%
2-[4-(Trifluoromethoxy)phenyl]-1,3-thiazolane
Hazard: Irritant
Formula:
C₁₀H₁₀F₃NOS MW: 249.26 g/mol Melting Pt: 78…80 °C |
MDL Number:
MFCD09027148 CAS Number: 937602-49-8 |
Specification Test Results
Melting point | 78-80°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...