N-[4-(1,3-Thiazolan-2-yl)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(1,3-Thiazolidin-2-yl)acetanilide

047752-500MG 047752-1G 047752-5G
101881-254EA 181.54 USD
101881-254 101881-256 101881-258
N-[4-(1,3-Thiazolan-2-yl)phenyl]acetamide ≥95%
N-[4-(1,3-Thiazolan-2-yl)phenyl]acetamide

Hazard: Irritant

Formula: C₁₁H₁₄N₂OS
MW: 222.31 g/mol
Melting Pt: 156…158 °C
MDL Number: MFCD09027147
CAS Number: 145300-45-4

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR