2-(4-Heptylphenyl)-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Hept-1-ylphenyl)-1,3-thiazolidine
047750-500MG
047750-1G
047750-5G
101881-242EA
181.54
USD
101881-242
101881-244
101881-246
2-(4-Heptylphenyl)-1,3-thiazolane ≥95%
2-(4-Heptylphenyl)-1,3-thiazolane
Hazard: Irritant
Formula:
C₁₆H₂₅NS Melting Pt: 67-69°C |
MDL Number:
MFCD09027145 CAS Number: 937602-48-7 |
Specification Test Results
Melting point | 67-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...