3-{[4-(Ethoxycarbonyl)piperidino]sulfonyl}benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

047748-500MG 047748-1G
101881-232EA 181.54 USD
101881-232 101881-234
3-{[4-(Ethoxycarbonyl)piperidino]sulfonyl}benzenecarboxylic acid ≥95%
3-{[4-(Ethoxycarbonyl)piperidino]sulfonyl}benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₆S
Melting Pt: 200…202 °C
MDL Number: MFCD09027143
CAS Number: 937604-24-5

Order Now

Specification Test Results

Melting point 200-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR