2-(4-Bromo-2-fluorophenyl)-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
047744-500MG
047744-1G
047744-5G
101881-218EA
181.54
USD
101881-218
101881-220
101881-222
2-(4-Bromo-2-fluorophenyl)-1,3-thiazolane ≥95%
2-(4-Bromo-2-fluorophenyl)-1,3-thiazolane
Hazard: Irritant
Formula:
C₉H₉BrFNS MW: 262.15 g/mol Melting Pt: 70…72 °C |
MDL Number:
MFCD09027142 CAS Number: 937602-46-5 |
Specification Test Results
Melting point | 70-72°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...