5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

047743-500MG 047743-1G 047743-5G
101881-212EA 556.92 USD
101881-212 101881-214 101881-216
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₄H₉ClF₃NO₃
Melting Pt: 218…220 °C
MDL Number: MFCD04038241
CAS Number: 886761-94-0

Order Now

Specification Test Results

Melting point 218-220°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR