2-(4-Ethylphenyl)-1,3-thiazolane ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(4-Ethylphenyl)-1,3-thiazolidine
047741-500MG
047741-1G
047741-5G
101881-200EA
181.54
USD
101881-200
101881-202
101881-204
2-(4-Ethylphenyl)-1,3-thiazolane ≥95%
2-(4-Ethylphenyl)-1,3-thiazolane
Hazard: Irritant
Formula:
C₁₁H₁₅NS Melting Pt: 91-93°C |
MDL Number:
MFCD09027140 CAS Number: 145300-46-5 |
Specification Test Results
Melting point | 91-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...