2-[4-(Benzyloxy)phenyl]-1,3-thiazolane ≥95%

Supplier: Matrix Scientific

047740-500MG 047740-1G 047740-5G
101881-194EA 138.46 USD
101881-194 101881-196 101881-198
2-[4-(Benzyloxy)phenyl]-1,3-thiazolane ≥95%
2-[4-(Benzyloxy)phenyl]-1,3-thiazolane

Hazard: Irritant

Formula: C₁₆H₁₇NOS
Melting Pt: 134…136 °C
MDL Number: MFCD09027139
CAS Number: 937602-44-3

Order Now

Specification Test Results

Melting point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR