3-(Piperidinosulfonyl)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Piperidin-1-ylsulphonyl)benzoic acid
047738-500MG
047738-1G
101881-184EA
181.54
USD
101881-184
101881-186
3-(Piperidinosulfonyl)benzenecarboxylic acid ≥95%
3-(Piperidinosulfonyl)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₅NO₄S MW: 269.32 g/mol Melting Pt: 179…181 °C |
MDL Number:
MFCD00760901 CAS Number: 7311-93-5 |
Specification Test Results
Melting point | 179-181°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...