2-[4-(tert-Butyl)phenyl]-1,3-thiazolane ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[4-(tert-Butyl)phenyl]-1,3-thiazolidine

047736-500MG 047736-1G 047736-5G
101881-174EA 181.54 USD
101881-174 101881-176 101881-178
2-[4-(tert-Butyl)phenyl]-1,3-thiazolane ≥95%
2-[4-(tert-Butyl)phenyl]-1,3-thiazolane

Hazard: Irritant

Formula: C₁₃H₁₉NS
MW: 221.37 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD09027137
CAS Number: 210625-01-7

Order Now

Specification Test Results

Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR