2-[(3-Methylbenzyl)sulfanyl]ethylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(3-Methylbenzyl)thio]ethylamine hydrochloride

047735-500MG 047735-1G 047735-5G
101881-168EA 181.54 USD
101881-168 101881-170 101881-172
2-[(3-Methylbenzyl)sulfanyl]ethylamine hydrochloride ≥95%
2-[(3-Methylbenzyl)sulfanyl]ethylamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆ClNS
MW: 217.76 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD09027136
CAS Number: 1171226-58-6

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR