2-Phenylthiazolidine ≥97%

Supplier: Matrix Scientific

047732-500MG 047732-1G 047732-5G
101881-150EA 181.54 USD
101881-150 101881-152 101881-154
2-Phenylthiazolidine ≥97%
2-Phenylthiazolidine

Hazard: Irritant

Formula: C₉H₁₁NS
MW: 165.254 g/mol
Melting Pt: 110…112 °C
MDL Number: MFCD00128238
CAS Number: 4569-82-8

Order Now

Specification Test Results

Melting point 110-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR