2-[(4-Fluorobenzyl)sulfanyl]ethylamine hydrochloride ≥95%
Supplier: Matrix Scientific
047731-500MG
047731-1G
047731-5G
101881-144EA
181.54
USD
101881-144
101881-146
101881-148
2-[(4-Fluorobenzyl)sulfanyl]ethylamine hydrochloride ≥95%
2-[(4-Fluorobenzyl)sulfanyl]ethylamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClFNS MW: 221.73 g/mol Melting Pt: 167…169 °C |
MDL Number:
MFCD09027133 CAS Number: 1170404-43-9 |
Specification Test Results
Melting point | 167-169°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...