Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
046635-500MG
046635-1G
101876-350EA
556.92
USD
101876-350
101876-352
Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate ≥95%
Methyl-5-chloro-2,6-dihydroxy-4-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₆H₅ClN₂O₄ Melting Pt: 262…263 °C |
MDL Number:
MFCD12025873 CAS Number: 91447-90-4 |
Specification Test Results
Melting point | 262-263°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...