6-Chloro-8-iodoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046630-500MG 046630-1G
101876-338EA 747.69 USD
101876-338 101876-340
6-Chloro-8-iodoimidazo[1,2-a]pyridine ≥95%
6-Chloro-8-iodoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄ClIN₂
Melting Pt: 142-143°C
MDL Number: MFCD10758078
CAS Number: 1033463-28-3

Order Now

Specification Test Results

Melting point 142-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR