6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid ≥95%
Supplier: Matrix Scientific
046629-500MG
046629-1G
101876-334EA
306.15
USD
101876-334
101876-336
6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid ≥95%
6-[4-(tert-Butoxycarbonyl)piperazino]nicotinic acid
Hazard: Irritant
Formula:
C₁₅H₂₁N₃O₄ MW: 307.35 g/mol Melting Pt: 228…229 °C |
MDL Number:
MFCD06245543 CAS Number: 201809-22-5 |
Specification Test Results
Melting point | 228-229°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...