2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2,3-Dihydro-7-azaindole
046626-1G
046626-5G
101876-326EA
15.38
USD
101876-326
101876-328
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine ≥95%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine
Hazard: Irritant
Formula:
C₇H₈N₂ MW: 120.15 g/mol Melting Pt: 80…82 °C |
MDL Number:
MFCD06659751 CAS Number: 10592-27-5 |
Specification Test Results
Melting point | 80-82°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...