4-Iodo-1H-pyrrole-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
046625-500MG
046625-1G
046625-5G
101876-320EA
700
USD
101876-320
101876-322
101876-324
4-Iodo-1H-pyrrole-2-carboxylic acid ≥95%
4-Iodo-1H-pyrrole-2-carboxylic acid
Hazard: Irritant
Formula:
C₅H₄INO₂ Melting Pt: 184…186 °C |
MDL Number:
MFCD10758077 CAS Number: 252861-26-0 |
Specification Test Results
Melting point | 184-186°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...