1-(4-Nitrophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

046622-500MG 046622-1G
101876-316EA 278.46 USD
101876-316 101876-318
1-(4-Nitrophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine ≥95%
1-(4-Nitrophenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Hazard: Irritant

Formula: C₁₁H₈N₆O₂
MW: 256.23 g/mol
Melting Pt: >250 °C
MDL Number: MFCD00128355
CAS Number: 65973-73-1

Order Now

Specification Test Results

Melting point >250°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR