6,8-Dichloro-3-iodoimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046621-500MG 046621-1G
101876-312EA 556.92 USD
101876-312 101876-314
6,8-Dichloro-3-iodoimidazo[1,2-a]pyridine ≥95%
6,8-Dichloro-3-iodoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₃Cl₂IN₂
Melting Pt: 147-150°C
MDL Number: MFCD10758076
CAS Number: 1033463-27-2

Order Now

Specification Test Results

Melting point 147-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR