3-[1-(4-Methyl-1,3-benzothiazol-2-yl)hydrazino]propanenitrile ≥95%

Supplier: Matrix Scientific

046620-500MG 046620-1G
101876-308EA 556.92 USD
101876-308 101876-310
3-[1-(4-Methyl-1,3-benzothiazol-2-yl)hydrazino]propanenitrile ≥95%
3-[1-(4-Methyl-1,3-benzothiazol-2-yl)hydrazino]propanenitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₄S
Melting Pt: 120…122 °C
MDL Number: MFCD10758075
CAS Number: 1033463-26-1

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR