3-[1-(1,3-Benzothiazol-2-yl)hydrazino]propanenitrile ≥95%

Supplier: Matrix Scientific

046619-500MG 046619-1G
101876-304EA 350.77 USD
101876-304 101876-306
3-[1-(1,3-Benzothiazol-2-yl)hydrazino]propanenitrile ≥95%
3-[1-(1,3-Benzothiazol-2-yl)hydrazino]propanenitrile

Hazard: Irritant

Formula: C₁₀H₁₀N₄S
MW: 218.28 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00099326
CAS Number: 86691-41-0

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR