5-Amino-1-(1,3-benzoxazol-2-yl)-2,3-dihydro-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

046616-500MG 046616-1G
101876-290EA 278.46 USD
101876-290 101876-292
5-Amino-1-(1,3-benzoxazol-2-yl)-2,3-dihydro-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1-(1,3-benzoxazol-2-yl)-2,3-dihydro-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₉N₅O
Melting Pt: >300 °C
MDL Number: MFCD10758073
CAS Number: 1033463-25-0

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR