4-Chloro-N-[3-(2-oxo-1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%

Supplier: Matrix Scientific

046615-500MG 046615-1G
101876-286EA 181.54 USD
101876-286 101876-288
4-Chloro-N-[3-(2-oxo-1-pyrrolidinyl)propyl]benzenesulfonamide ≥95%
4-Chloro-N-[3-(2-oxo-1-pyrrolidinyl)propyl]benzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₃S
Melting Pt: 122…124 °C
MDL Number: MFCD01185588
CAS Number: 321713-50-2

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR