Methyl-4-(acetylamino)-2-[(4-nitrobenzyl)oxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
046614-500MG
046614-1G
046614-5G
101876-280EA
556.92
USD
101876-280
101876-282
101876-284
Methyl-4-(acetylamino)-2-[(4-nitrobenzyl)oxy]benzenecarboxylate ≥95%
Methyl-4-(acetylamino)-2-[(4-nitrobenzyl)oxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₇H₁₆N₂O₆ Melting Pt: 170…171 °C |
MDL Number:
MFCD10758072 CAS Number: 1033463-24-9 |
Specification Test Results
Melting point | 170-171°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...