Methyl-4-(2-formylphenoxy)-3-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
046610-500MG
046610-1G
046610-5G
101876-260EA
278.46
USD
101876-260
101876-262
101876-264
Methyl-4-(2-formylphenoxy)-3-nitrobenzenecarboxylate ≥95%
Methyl-4-(2-formylphenoxy)-3-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₁NO₆ Melting Pt: 117…119 °C |
MDL Number:
MFCD10758070 CAS Number: 80248-06-2 |
Specification Test Results
Melting point | 117-119°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...