4-Amino-3-(4-bromophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Bromophenyl)-1H-pyrazol-4-amine

046605-500MG 046605-1G
101876-244EA 607.69 USD
101876-244 101876-246
4-Amino-3-(4-bromophenyl)-1H-pyrazole ≥95%
4-Amino-3-(4-bromophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₈BrN₃
Melting Pt: 165…167 °C
MDL Number: MFCD10758068
CAS Number: 91857-98-6

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR