3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046599-500MG 046599-1G
101876-226EA 218.46 USD
101876-226 101876-228
3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine ≥95%
3-Bromo-6,8-dichloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₃BrCl₂N₂
Melting Pt: 111…113 °C
MDL Number: MFCD09607939
CAS Number: 952183-48-1

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR