4-Methyl-1H-indazol-7-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Amino-4-methyl-1H-indazole
046594-500MG
046594-1G
101876-206EA
747.69
USD
101876-206
101876-208
4-Methyl-1H-indazol-7-amine ≥95%
4-Methyl-1H-indazol-7-amine
Hazard: Irritant
Formula:
C₈H₉N₃ MW: 147.18 g/mol Melting Pt: 211…214 °C |
MDL Number:
MFCD09607935 CAS Number: 952183-44-7 |
Specification Test Results
Melting point | 211-214°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...