tert-Butyl-4-[4-methyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[4-Methyl-5-(methylthio)-4H-1,2,4-triazol-3-yl]piperidine, N-BOC protected

046593-500MG 046593-1G 046593-5G
101876-200EA 556.92 USD
101876-200 101876-202 101876-204
tert-Butyl-4-[4-methyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate ≥95%
tert-Butyl-4-[4-methyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]tetrahydro-1(2H)-pyridine carboxylate

Hazard: Irritant

Formula: C₁₄H₂₄N₄O₂S
MW: 312.44 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD09607934
CAS Number: 952183-43-6

Order Now

Specification Test Results

Melting point 116-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR