Thieno[3,2-b]thiophene-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
046592-500MG
046592-1G
046592-5G
101876-194EA
556.92
USD
101876-194
101876-196
101876-198
Thieno[3,2-b]thiophene-2-carbohydrazide ≥95%
Thieno[3,2-b]thiophene-2-carbohydrazide
Hazard: Irritant
Formula:
C₇H₆N₂OS₂ Melting Pt: 188…190 °C |
MDL Number:
MFCD04112097 CAS Number: 685114-87-8 |
Specification Test Results
Melting point | 188-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...