6,8-Dichloroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

046588-500MG 046588-1G 046588-5G
101876-174EA 110.77 USD
101876-174 101876-176 101876-178
6,8-Dichloroimidazo[1,2-a]pyridine ≥95%
6,8-Dichloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
Melting Pt: 152-154°C
MDL Number: MFCD06761692
CAS Number: 858516-69-5

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR