4-Chloro-1H-indazole ≥95%

Supplier: Matrix Scientific

046587-500MG 046587-1G 046587-5G
101876-168EA 375.38 USD
101876-168 101876-170 101876-172
4-Chloro-1H-indazole ≥95%
4-Chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 155…157 °C
MDL Number: MFCD00211065
CAS Number: 13096-96-3

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR