7-Bromo-1-methyl-5-nitro-1H-indazole ≥95%

Supplier: Matrix Scientific

046586-500MG 046586-1G
101876-164EA 747.69 USD
101876-164 101876-166
7-Bromo-1-methyl-5-nitro-1H-indazole ≥95%
7-Bromo-1-methyl-5-nitro-1H-indazole

Hazard: Irritant

Formula: C₈H₆BrN₃O₂
Melting Pt: 166…168 °C
MDL Number: MFCD09607926
CAS Number: 952183-39-0

Order Now

Specification Test Results

Melting point 166-168°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR