6-Bromo-1H-indazole ≥95%
Supplier: Matrix Scientific
046585-1G
046585-5G
046585-25G
101876-158EA
15.38
USD
101876-158
101876-160
101876-162
6-Bromo-1H-indazole ≥95%
6-Bromo-1H-indazole
Hazard: Irritant
Formula:
C₇H₅BrN₂ MW: 197.0300 g/mol Melting Pt: 180…182 °C |
MDL Number:
MFCD03265457 CAS Number: 79762-54-2 |
Specification Test Results
Melting point | 180-182°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...