1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione ≥95%
Supplier: Matrix Scientific
046584-500MG
046584-1G
046584-5G
101876-152EA
200
USD
101876-152
101876-154
101876-156
1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione ≥95%
1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione
Hazard: Irritant
Formula:
C₁₀H₈F₃N₃O₃ Melting Pt: 171…174 °C |
MDL Number:
MFCD09607925 CAS Number: 952183-38-9 |
Specification Test Results
Melting point | 171-174°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...