2-Methyl-2H-indazol-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-methyl-2H-indazole

046583-500MG 046583-1G
101876-148EA 170.77 USD
101876-148 101876-150
2-Methyl-2H-indazol-6-amine ≥95%
2-Methyl-2H-indazol-6-amine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD00464866
CAS Number: 50593-30-1

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR