1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046582-500MG 046582-1G
101876-144EA 556.92 USD
101876-144 101876-146
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
1-(2-Chloro-6-fluorobenzyl)-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₉ClFNO₂
MW: 265.67 g/mol
Melting Pt: 130…131 °C
MDL Number: MFCD09817431
CAS Number: 952183-37-8

Order Now

Specification Test Results

Melting point 130-131°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR