5-Iodo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

046581-500MG 046581-1G
101876-140EA 556.92 USD
101876-140 101876-142
5-Iodo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Iodo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Melting Pt: 228…230 °C
MDL Number: MFCD09607924
CAS Number: 952183-36-7

Order Now

Specification Test Results

Melting point 228-230°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR