5-Chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
046579-500MG
046579-1G
101876-132EA
556.92
USD
101876-132
101876-134
5-Chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Chloro-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₆H₄ClNO₂ MW: 157.56 g/mol Melting Pt: 255…257 °C |
MDL Number:
MFCD09607922 CAS Number: 627501-18-2 |
Specification Test Results
Melting point | 255-257°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...