5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
046578-500MG
046578-1G
101876-128EA
600
USD
101876-128
101876-130
5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde ≥95%
5-Bromo-6-oxo-1,6-dihydro-3-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 232…234 °C |
MDL Number:
MFCD09607921 CAS Number: 952183-35-6 |
Specification Test Results
Melting point | 232-234°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...