2-Methyl-4-thiazolecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Methylthiazole-4-carboxylic acid
046577-1G
046577-5G
046577-25G
101876-124EA
138.46
USD
101876-124
101876-126
102551-348
2-Methyl-4-thiazolecarboxylic acid ≥95%
2-Methyl-4-thiazolecarboxylic acid
Hazard: Irritant
Formula:
C₅H₅NO₂S MW: 143.1637 g/mol Melting Pt: 146-148°C |
MDL Number:
MFCD03407332 CAS Number: 35272-15-2 |
Specification Test Results
Melting point | 146-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...