N-(2-Hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
046574-500MG
046574-1G
046574-5G
101876-106EA
121.54
USD
101876-106
101876-108
101876-110
N-(2-Hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide ≥95%
N-(2-Hydroxyethyl)-4-(1H-imidazol-1-yl)benzenecarboxamide
Hazard: Irritant
Formula:
C₁₂H₁₃N₃O₂ MW: 231.26 g/mol Melting Pt: 175…177 °C |
MDL Number:
MFCD09607920 CAS Number: 952183-34-5 |
Specification Test Results
Melting point | 175-177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...